Czech Republic
Order Entry
Chicken native albumin (from egg)
Chicken native albumin (from egg)
Supplier:  Merck
Certificates
Chicken native albumin (from egg)
Supplier:  Merck

Specifications

  • CAS No.:
    9006-59-1
  • Protein/peptide type:
    Native
  • Source:
    Egg
  • Species:
    Chicken
  • Storage conditions:
    Refrigerator
  • Protein/peptide name:
    Albumin

Specifications

Product Information

  • Catalog #
    Supplier #
    Pack type
    Pk
    Availability
    Price
  • SIALA2512-250MG
    A2512-250MG
    Glass bottle for solids
    250 mg
       
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 2 500,00 Kč
    2 500,00 Kč
    1 * 250 mg
       2 500,00 Kč

     : A2512-250MG
     : SIALA2512-250MG
  • SIALA2512-1G
    A2512-1G
    Glass bottle for solids
    1 g
       
       
    1 * 1 g
    1 * 1 g - 6 900,00 Kč
    6 900,00 Kč
    1 * 1 g
       6 900,00 Kč

     : A2512-1G
     : SIALA2512-1G
  • SIALA2512-5G
    A2512-5G
    Glass bottle for solids
    5 g
       
       
    1 * 5 g
    1 * 5 g - 23 640,00 Kč
    23 640,00 Kč
    1 * 5 g
       23 640,00 Kč

     : A2512-5G
     : SIALA2512-5G