Tisk…

Polyethylene glycol 40,000, Supelco®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

96699-1G-FEA 8310 CZK
96699-1G-F
Polyethylene glycol 40,000, Supelco®
Polyethylene glycol 40,000
Vzorec: H(OCH₂CH₂)nOH
MW: 40000 g/mol
Storage Temperature: Ambient
CAS číslo: 25322-68-3
EINECS: 500-038-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR