Tisk…

(±)-2-Phenylbutyric acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIALP20955-100GEA 1460 CZK
SIALP20955-100G
(±)-2-Phenylbutyric acid, Sigma-Aldrich®
(±)-2-Phenylbutyric acid
Vzorec: C₁₀H₁₂O₂
MW: 164,2 g/mol
Bod varu: 270 °C (1013 hPa)
Bod tání: 39…42 °C
MDL Number: MFCD00002667
CAS číslo: 90-27-7
EINECS: 201-982-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR