(±)-1-Hexen-3-ol, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIALH12854-10GEA
1870
CZK
SIALH12854-10G
(±)-1-Hexen-3-ol, Sigma-Aldrich®
(±)-1-Hexen-3-ol
Vzorec:
C₆H₁₂O MW: 100,16 g/mol Bod varu: 134…135 °C (1013 hPa) Bod tání: –72 °C Hustota: 0,884 g/cm³ (20 °C) |
MDL Number:
MFCD00004581 CAS číslo: 4798-44-1 EINECS: 225-355-0 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...