Paladium 5% on barium sulphate hydrogenation catalyst (oxidised form), unreduced
Dodavatel: Thermo Scientific
ACRO195060500EA
20700
CZK
ACRO195060500
ACRO195060100
Paladium 5% on barium sulphate hydrogenation catalyst (oxidised form), unreduced
Palladium
Vzorec:
Pd MW: 106,420g,mol Storage Temperature: Ambient |
MDL Number:
MFCD03613605 CAS číslo: 7440-05-3 EINECS: 231-115-6 |
Výsledky specifikačního testu
Appearance | Brown Powder |
Assay | 4.50 to 5.50 % (Pd) |
Water | ≤5.0 % (K.F.) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...