Tisk…
ESD safety shoes, lace-up, uvex business casual, 9510
Dodavatel: Uvex
uvex uvex business casual
UVEX9510.8/47EA 3070 CZK
UVEX9510.8/47 UVEX9510.8/46 UVEX9510.8/38 UVEX9510.8/48 UVEX9510.8/39 UVEX9510.8/41 UVEX9510.8/40 UVEX9510.8/43 UVEX9510.8/42
ESD safety shoes, lace-up, uvex business casual, 9510
Obuv
Lightweight, casual business shoes made from breathable leather upper, soft padding on collar and tongue plus a self-cleaning, lightweight DUO PU sole resistant to oil and petrol, short-term heat resistance up to 120 °C.
  • Climazone technology for improved comfort
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Anti-slip sole with shock absorption at heel and front of the foot
  • Slip and abrasion resistant plus shock-absorbent sole
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Colour: Black
Width: 11
Certifikace: EN 61340 (ESD); EN ISO 20345:2011; S1, SRC