Tisk…
ESD safety shoes, sandals, Velcro®, uvex motion light, 6980
Dodavatel: Uvex
uvex motion light
UVEX6980.8/43EA 2170 CZK
UVEX6980.8/43 UVEX6980.8/42 UVEX6980.8/40 UVEX6980.8/45 UVEX6980.8/38 UVEX6980.8/41 UVEX6980.8/46 UVEX6980.8/47 UVEX6980.8/44 UVEX6980.8/39 UVEX6980.8/48
ESD safety shoes, sandals, Velcro®, uvex motion light, 6980
Obuv
Ultralight ESD sandals made from nubuck leather, soft padded collar and tongue, breathable distance mesh and an extremely flexible dual density PUR sole (slip-, abrasion- and cut-resistant, resistant to oil and petrol and withstands brief exposure to heat of approx. +120 °C). Free of lacquer wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Anti-twist heel cap for better stability and protection from ankle twisting
  • Ergonomically designed, self-cleaning tread
  • Protective steel toecap (200 J)
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Colour: Black
Width: 11
Certifikace: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S1, SRC