Ethyl acetate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Acetic acid ethyl ester
:141-78-6
:
:CH₃COOC₂H₅
:MFCD00009171
:88.11 g/mol
:13,03792
:77.1 °C (1013 hPa)
:–83 °C
:0.902 g/cm³ (20 °C)
:205-500-4
:1173
:3,II
:M-Clarity™ quality level = MQ300
:–4 °C
:01-2119475103-46
:GHS02
:GHS07
:Ambient
Specifications
- Environmentally Preferable:
Specifications
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- SIALW241415-1KG-KW241415-1KG-KGlass bottle for solids1 kg1 * 1 kg3 540,00 Kč
- SIALW241415-5KG-KW241415-5KG-KGlass bottle for solids5 kg1 * 5 kg7 330,00 Kč
- SIALW241415-10KG-KW241415-10KG-KGlass bottle for solids10 kg1 * 10 kg8 850,00 Kč