Tisk…

Methyl valerate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL148997-100MLEA 1220 CZK
SIAL148997-100ML
Methyl valerate, Sigma-Aldrich®
Methyl valerate
Vzorec: C₆H₁₂O₂
MW: 116,16 g/mol
Bod varu: 126,5 °C (1013 hPa)
Bod tání: –91 °C
Hustota: 0,89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009478
CAS číslo: 624-24-8
EINECS: 210-838-0
UN: 3272
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR