This mini hotplate-stirrer is a rugged, compact unit that can heat and stir up to 1000 ml of liquid. Ceramic top features reflective white, chemical resistant top plate that is easy to clean. Bi-metallic thermostat offers reliable temperature control.
- Compact design saves bench space
- Ideal for educational laboratories
- Powerful motor and magnet deliver reliable and consistent stirring
- Powerful heater reaches maximum temperature in only minutes (boil 300 ml of water in 18 minutes)
- Adjustment knobs to control speed and temperature
- Can be used at 5 to 40 °C (from 20 to max. 80% relative humidity, non condensing)
Built-in support rod holder with locking knob accepts optional support rod and clamp kit.
Informace pro objednávání: Supplied with a 38 mm PTFE coated stirring bar, 3-wire cord and plug.