Glycin, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
SIAL50058-1KGEA
6530
CZK
SIAL50058-1KG
Glycin, Sigma-Aldrich®
Glycin
Vzorec:
NH₂CH₂COOH MW: 75,07 g/mol Bod varu: 181 °C (1013 hPa) Bod tání: 83 °C Hustota: 1,64 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008131 CAS číslo: 56-40-6 EINECS: 200-272-2 Merck Index: 13,04504 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...