Tisk…

(2,3,6-Trifluorophenyl)acetic acid, Sigma-Aldrich®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ200

SIAL446572-1GEA 4660 CZK
SIAL446572-1G
(2,3,6-Trifluorophenyl)acetic acid, Sigma-Aldrich®
(2,3,6-Trifluorophenyl)acetic acid
Vzorec: C₆H₂F₃CH₂COOH
MW: 190,12 g/mol
MDL Number: MFCD00061217
CAS číslo: 114152-23-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR