Tisk…

2-Heptyn-1-ol, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL630810-5GEA 2850 CZK
SIAL630810-5G
2-Heptyn-1-ol, Sigma-Aldrich®
2-Heptyn-1-ol
Vzorec: C₇H₁₂O
MW: 112,17 g/mol
MDL Number: MFCD00039541
CAS číslo: 1002-36-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR