4-Chloro-2-methoxybenzoic acid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL145793-5GEA
1450
CZK
SIAL145793-5G
4-Chloro-2-methoxybenzoic acid, Sigma-Aldrich®
4-Chloro-2-methoxybenzoic acid
Vzorec:
C₈H₇ClO₃ MW: 186,59 g/mol Bod varu: 308 °C (1013 hPa) Bod tání: 148 °C |
MDL Number:
MFCD00002532 CAS číslo: 57479-70-6 EINECS: 260-761-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...