Tisk…

Terephthalonitrile, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIALD76722-25GEA 1110 CZK
SIALD76722-25G
Terephthalonitrile, Sigma-Aldrich®
Terephthalonitrile
Vzorec: C₈H₄N₂
MW: 128,13 g/mol
Bod tání: 223…226 °C
Hustota: 1,29 g/cm³ (20 °C)
MDL Number: MFCD00001810
CAS číslo: 623-26-7
EINECS: 210-783-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR