3,5-Dimethoxyphenyl isocyanate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL478164-2GEA
3290
CZK
SIAL478164-2G
3,5-Dimethoxyphenyl isocyanate, Sigma-Aldrich®
3,5-Dimethoxyphenyl isocyanate
Vzorec:
C₉H₉NO₃ MW: 179,18 g/mol Storage Temperature: Refrigerator |
CAS číslo:
54132-76-2 UN: 3335 ADR: 9, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...