Tisk…

2-Bromobenzyl alcohol

Dodavatel: Molekula

MOLEM25429515EA 0 CZK
MOLEM25429515
2-Bromobenzyl alcohol
2-Bromobenzyl alcohol
Vzorec: C₇H₇BrO
MW: 187,04 g/mol
Bod varu: 255 °C (1013 hPa)
Bod tání: 80…82 °C
MDL Number: MFCD00004600
CAS číslo: 18982-54-2
EINECS: 242-719-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR