2,5-Dichlorobenzyl alcohol, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
SIAL183601-10GEA
5530
CZK
SIAL183601-10G
2,5-Dichlorobenzyl alcohol, Sigma-Aldrich®
2,5-Dichlorobenzyl alcohol
Vzorec:
C₇H₆Cl₂O MW: 177,03 g/mol Bod varu: 264 °C (1013 hPa) Bod tání: 80 °C |
MDL Number:
MFCD00004607 CAS číslo: 34145-05-6 EINECS: 251-850-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...