2',5'-Difluoroacetophenone, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL264261-1GEA
1250
CZK
SIAL264261-1G
2',5'-Difluoroacetophenone, Sigma-Aldrich®
2',5'-Difluoroacetophenone
Vzorec:
C₈H₆F₂O MW: 156,13 g/mol Bod varu: 70…80 °C (15 mmHg) Hustota: 1,2099 g/cm³ (22 °C) |
MDL Number:
MFCD00009898 CAS číslo: 1979-36-8 EINECS: 217-837-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...