Tisk…

2-Ethylphenyl isocyanate ≥99%

Dodavatel: Thermo Scientific
Nebezpečí

L15252.06EA 2670 CZK
L15252.06
2-Ethylphenyl isocyanate ≥99%
2-Ethylphenyl isocyanate
Vzorec: C₉H₉NO
MW: 147,18 g/mol
Bod varu: 56 °C (3 mmHg)
Hustota: 1,043
Bod vzplanutí: 78 °C (172 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00002013
CAS číslo: 40411-25-4
EINECS: 000-000-0
UN: 2206
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR