Tisk…

4-Dodecylaniline, Sigma-Aldrich®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ100

SIAL233544-5GEA 0 CZK
SIAL233544-5G
4-Dodecylaniline, Sigma-Aldrich®
4-Dodecylaniline
Vzorec: C₁₈H₃₁N
MW: 261,45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007919
CAS číslo: 104-42-7
EINECS: 203-201-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR