Tisk…

N-Ethyl-m-toluidine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIAL04930-500MLEA 5870 CZK
SIAL04930-500ML
N-Ethyl-m-toluidine, Sigma-Aldrich®
N-Ethyl-m-toluidine
Vzorec: C₉H₁₃N
MW: 135,21 g/mol
Bod varu: 221 °C (1013 hPa)
Bod tání: 9 °C
Hustota: 0,9264 g/cm³ (15 °C)
MDL Number: MFCD00009027
CAS číslo: 102-27-2
EINECS: 203-019-4
UN: 2754
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR