Tisk…

N-Methyl-O-toluidine, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

SIAL467278-25MLEA 0 CZK
SIAL467278-25ML
N-Methyl-O-toluidine, Sigma-Aldrich®
N-Methyl-O-toluidine
Vzorec: C₈H₁₁N
MW: 121,18 g/mol
Bod varu: 206…208 °C (1013 hPa)
Hustota: 0,973 g/cm³ (15 °C)
MDL Number: MFCD00025627
CAS číslo: 611-21-2
EINECS: 210-260-9
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR