Tisk…

N-Methyl-p-toluidine, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ100

SIAL494208-25MLEA 3830 CZK
SIAL494208-25ML
N-Methyl-p-toluidine, Sigma-Aldrich®
N-Methyl-p-toluidine
Vzorec: C₈H₁₁N
MW: 121,18 g/mol
Bod varu: 89 °C (1013 hPa)
Hustota: 0,9625 g/cm³ (20 °C)
MDL Number: MFCD00025628
CAS číslo: 623-08-5
EINECS: 210-769-6
UN: 2810
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR