Tisk…

2-Nitrobenzyl alcohol

MOLEM83431071EA 0 CZK
MOLEM83431071
2-Nitrobenzyl alcohol
2-Nitrobenzyl alcohol
Vzorec: C₇H₇NO₃
MW: 153,14 g/mol
Bod varu: 270 °C (1013 hPa)
Bod tání: 71…72 °C
MDL Number: MFCD00007186
CAS číslo: 612-25-9
EINECS: 210-302-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR