5-Nitro-o-cresol
|
Varování
|
MOLEM46352676EA
0
CZK
MOLEM46352676
5-Nitro-o-cresol
5-Nitro-o-cresol
Vzorec:
C₇H₇NO₃ MW: 153,14 g/mol |
MDL Number:
MFCD00043909 CAS číslo: 5428-54-6 EINECS: 226-580-7 UN: 2446 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...