Tisk…

2-Isopropylanisole, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL550736-25GEA 4310 CZK
SIAL550736-25G
2-Isopropylanisole, Sigma-Aldrich®
2-Isopropylanisole
Vzorec: C₁₀H₁₄O
MW: 150,22 g/mol
MDL Number: MFCD00798590
CAS číslo: 2944-47-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR