Tisk…

3-Toluenethiol, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIALT28517-5GEA 2620 CZK
SIALT28517-5G
3-Toluenethiol, Sigma-Aldrich®
3-Toluenethiol
Vzorec: C₇H₈S
MW: 124,21 g/mol
Bod varu: 195 °C (1013 hPa)
Hustota: 1,06251 g/cm³ (0 °C)
MDL Number: MFCD00004843
CAS číslo: 108-40-7
EINECS: 203-578-4
UN: 2810
ADR: 6.1,III
Merck Index: 12,09461

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR