Tisk…

Tridecylbenzene, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL113247-5GEA 3240 CZK
SIAL113247-5G
Tridecylbenzene, Sigma-Aldrich®
Tridecylbenzene
Vzorec: C₁₉H₃₂
MW: 260,46 g/mol
Bod varu: 341 °C (1013 hPa)
Bod tání: 10 °C
Hustota: 0,855 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008980
CAS číslo: 123-02-4
EINECS: 204-592-3
UN: 3082
ADR: 9,III
Merck Index: 12,09792

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR