Tisk…

2,4-Dimethylbenzoic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL138169-10GEA 1760 CZK
SIAL138169-10G
2,4-Dimethylbenzoic acid, Sigma-Aldrich®
2,4-Dimethylbenzoic acid
Vzorec: C₉H₁₀O₂
MW: 150,18 g/mol
Bod varu: 267 °C (1013 hPa)
Bod tání: 123…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00002480
CAS číslo: 611-01-8
EINECS: 210-246-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR