Tisk…

2-Fluoro-m-xylene, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL452866-5MLEA 0 CZK
SIAL452866-5ML
2-Fluoro-m-xylene, Sigma-Aldrich®
2-Fluoro-m-xylene
Vzorec: C₈H₉F
MW: 124,16 g/mol
Bod varu: 165…167 °C (1013 hPa)
Hustota: 0,988 g/cm³ (20 °C)
MDL Number: MFCD00039215
CAS číslo: 443-88-9
EINECS: 207-144-5
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR