Tisk…

3'-(Trifluoromethyl)propiophenone, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL538698-5GEA 3570 CZK
SIAL538698-5G
3'-(Trifluoromethyl)propiophenone, Sigma-Aldrich®
3'-(Trifluoromethyl)propiophenone
Vzorec: C₁₀H₉F₃O
MW: 202,18 g/mol
MDL Number: MFCD00039227
CAS číslo: 1533-03-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR