Tisk…

2,6-Bis(trifluoromethyl)benzoic acid

MOLE11880383-5GEA 0 CZK
MOLE11880383-5G
2,6-Bis(trifluoromethyl)benzoic acid
2,6-Bis(trifluoromethyl)benzoic acid
Vzorec: C₉H₄F₆O₂
MW: 258,12 g/mol
Bod varu: 260 °C (1013 hPa)
Bod tání: 138…139 °C
MDL Number: MFCD00000376
CAS číslo: 24821-22-5
EINECS: 246-479-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR