Tisk…

2-Bromo-1,3,5-triisopropylbenzene, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

SIAL637939-5GEA 1060 CZK
SIAL637939-5G
2-Bromo-1,3,5-triisopropylbenzene, Sigma-Aldrich®
2-Bromo-1,3,5-triisopropylbenzene
Vzorec: C₁₅H₂₃Br
MW: 283,25 g/mol
MDL Number: MFCD00051547
CAS číslo: 21524-34-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR