Tisk…

3-Bromobiphenyl (PBB No. 2)

MOLEM58329326EA 0 CZK
MOLEM58329326
3-Bromobiphenyl (PBB No. 2)
3-Bromobiphenyl (PBB No. 2)
Vzorec: C₁₂H₉Br
MW: 233,11 g/mol
Bod varu: 299 °C (1013 hPa)
Bod tání: 9 °C
Hustota: 1,41 g/cm³ (20 °C)
MDL Number: MFCD00000082
CAS číslo: 2113-57-7
EINECS: 218-304-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR