Tisk…

2,3,3',5'-Tetrachlorobiphenyl (PCB No. 58)

Dodavatel: LGC Standards PROMOCHEM

PROOCIL-PCB-58-CEA 21370 CZK
PROOCIL-PCB-58-C
2,3,3',5'-Tetrachlorobiphenyl (PCB No. 58)
2,3,3',5'-Tetrachlorobiphenyl (PCB No. 58)
Vzorec: C₁₂H₆Cl₄
MW: 291,99 g/mol
CAS číslo: 41464-49-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR