Tisk…

3,3',4,4'-Tetrachlorobiphenyl (PCB No. 77)

PROOCIL-PCB-77-CEA 0 CZK
PROOCIL-PCB-77-C
3,3',4,4'-Tetrachlorobiphenyl (PCB No. 77)
3,3',4,4'-Tetrachlorobiphenyl (PCB No. 77)
Vzorec: C₁₂H₆Cl₄
MW: 291,99 g/mol
Bod varu: 381 °C (1013 hPa)
Bod tání: 173 °C
CAS číslo: 32598-13-3
UN: 3432
ADR: 9,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR