Tisk…

3,4-Dichlorobiphenyl (PCB No. 12)

PROOCIL-PCB-12-CEA 0 CZK
PROOCIL-PCB-12-C
3,4-Dichlorobiphenyl (PCB No. 12)
3,4-Dichlorobiphenyl (PCB No. 12)
Vzorec: C₁₂H₈Cl₂
MW: 223,1 g/mol
CAS číslo: 2974-92-7
UN: 2315
ADR: 9,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR