Tisk…

2,2',3,5',6-Pentachlorobiphenyl (PCB No. 95)

Dodavatel: LGC Standards PROMOCHEM

PROOCIL-PCB-95-CEA 21370 CZK
PROOCIL-PCB-95-C
2,2',3,5',6-Pentachlorobiphenyl (PCB No. 95)
2,2',3,5',6-Pentachlorobiphenyl (PCB No. 95)
Vzorec: C₁₂H₅Cl₅
MW: 326,44 g/mol
CAS číslo: 38379-99-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR