Tisk…

2,2',4,5',6-Pentachlorobiphenyl (PCB No. 103)

PROOCIL-PCB-103-CEA 0 CZK
PROOCIL-PCB-103-C
2,2',4,5',6-Pentachlorobiphenyl (PCB No. 103)
2,2',4,5',6-Pentachlorobiphenyl (PCB No. 103)
Vzorec: C₁₂H₅Cl₅
MW: 326,44 g/mol
CAS číslo: 60145-21-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR