Tisk…

2,3',5'-Trichlorobiphenyl (PCB No. 34)

Dodavatel: LGC Standards PROMOCHEM

PROOCIL-PCB-34-CEA 21370 CZK
PROOCIL-PCB-34-C
2,3',5'-Trichlorobiphenyl (PCB No. 34)
2,3',5'-Trichlorobiphenyl (PCB No. 34)
Vzorec: C₁₂H₇Cl₃
MW: 257,55 g/mol
CAS číslo: 37680-68-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR