Tisk…

2,3,3',4',6-Pentachlorobiphenyl (PCB No. 110)

Dodavatel: EHRENSTORFER GMBH

EHERC20011000EA 3800 CZK
EHERC20011000
2,3,3',4',6-Pentachlorobiphenyl (PCB No. 110)
2,3,3',4',6-Pentachlorobiphenyl (PCB No. 110)
Vzorec: C₁₂H₅Cl₅
MW: 326,44 g/mol
CAS číslo: 38380-03-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR