Tisk…
2,3,3',4,5'-Pentachlorobiphenyl (PCB No. 108)
Dodavatel: LGC Standards PROMOCHEM
PROOCIL-PCB-108-CEA 21370 CZK
PROOCIL-PCB-108-C
2,3,3',4,5'-Pentachlorobiphenyl (PCB No. 108)
2,3,3',4,5'-Pentachlorobiphenyl (PCB No. 108)
Vzorec: C₁₂H₅Cl₅
MW: 326,44 g/mol
CAS číslo: 70362-41-3