Tisk…

2-Chlorobiphenyl (PCB No. 1), Supelco®


M-Clarity™ quality level = MQ100

35586-100MGEA 0 CZK
35586-100MG
2-Chlorobiphenyl (PCB No. 1), Supelco®
2-Chlorobiphenyl (PCB No. 1)
Vzorec: C₁₂H₉Cl
MW: 188,66 g/mol
Bod varu: 274 °C (1013 hPa)
Bod tání: 34 °C
Hustota: 1,1499 g/cm³ (32 °C)
CAS číslo: 2051-60-7
EINECS: 218-125-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR