Tisk…

2,2',3,4,4',5,5',6-Octachlorobiphenyl (PCB No. 203)

Dodavatel: LGC Standards PROMOCHEM

PROOCIL-PCB-203-CEA 21370 CZK
PROOCIL-PCB-203-C
2,2',3,4,4',5,5',6-Octachlorobiphenyl (PCB No. 203)
2,2',3,4,4',5,5',6-Octachlorobiphenyl (PCB No. 203)
Vzorec: C₁₂H₂Cl₈
MW: 429,77 g/mol
CAS číslo: 52663-76-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR