Tisk…

2-Iodobiphenyl, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL529982-25GEA 16010 CZK
SIAL529982-25G
2-Iodobiphenyl, Sigma-Aldrich®
2-Iodobiphenyl
Vzorec: C₁₂H₉I
MW: 280,11 g/mol
MDL Number: MFCD00039396
CAS číslo: 2113-51-1
EINECS: 218-303-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR