p-Terphenyl, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
SIAL86470-50GEA
2570
CZK
SIAL86470-50G
p-Terphenyl, Sigma-Aldrich®
p-Terphenyl
Vzorec:
C₁₈H₁₄ MW: 230,31 g/mol Bod varu: 383 °C (1013 hPa) Bod tání: 210…211 °C Hustota: 1,234 g/cm³ (0 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003061 CAS číslo: 92-94-4 EINECS: 202-205-2 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...