Tisk…

5-Methylquinoxaline

Varování

MOLE10923708-5GEA 7910 CZK
MOLE10923708-5G
5-Methylquinoxaline
5-Methylquinoxaline
Vzorec: C₉H₈N₂
MW: 144,18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00012335
CAS číslo: 13708-12-8
EINECS: 237-246-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR