Tisk…

γ-Terpinene

Nebezpečí

EHERC17322325EA 0 CZK
EHERC17322325
γ-Terpinene
γ-Terpinene
Vzorec: C₁₀H₁₆
MW: 136,24 g/mol
Bod varu: 182 °C (1013 hPa)
Hustota: 0,846 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001537
CAS číslo: 99-85-4
EINECS: 202-794-6
UN: 2319
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR