Tisk…

2-Acetylcyclohexanone, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIAL179760-25GEA 4280 CZK
SIAL179760-25G SIAL179760-5G
2-Acetylcyclohexanone, Sigma-Aldrich®
2-Acetylcyclohexanone
Vzorec: C₈H₁₂O₂
MW: 140,18 g/mol
Hustota: 1,063 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001633
CAS číslo: 874-23-7
EINECS: 212-858-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR